HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Hexamethyldisilazane

120580100 120581000 120585000 120580025
ACRO120580100EA 22.9 EUR
ACRO120580100 ACRO120581000 ACRO120585000 ACRO120580025
HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98%
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
Formula: (CH₃)₃SiNHSi(CH₃)₃
MW: 161.39 g/mol
Boiling Pt: 125 °C (760 mmHg)
Melting Pt: –78 °C
Density: 0.76 g/cm³
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008259
CAS Number: 999-97-3
EINECS: 213-668-5
UN: 3286
ADR: 3 II
REACH: 01-2119438176-38
Merck Index: 13,04708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4090 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤30 APHA

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR