Calcium chloride dihydrate 77 - 80% 77-80, flake, extra_pure Drying

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

123350025 123350100
ACRO123350025EA 155 EUR
ACRO123350025 ACRO123350100
Calcium chloride dihydrate 77 - 80% 77-80, flake, extra_pure Drying
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600
Melting Pt: 175 °C
Density: 0.83 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

Specification Test Results

Appearance White to light grey Flakes
Titration Complexometric 77 to 80 % (CaCl2)
Melting point 81 to 84 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR