Diethyl (2-oxopropyl)phosphonate 96%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Diethyl acetylmethylphosphonate

123690050 123690250
ACRO123690050EA 75.6 EUR
ACRO123690050 ACRO123690250
Diethyl (2-oxopropyl)phosphonate 96%
Diethyl (2-oxopropyl)phosphonate
Formula: C₇H₁₅O₄P
MW: 194.17 g/mol
Boiling Pt: 98 °C (1 mmHg)
Density: 1.11 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00044728
CAS Number: 1067-71-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4325 to 1.4355 (20°C, 589 nm)
GC ≥95 %
Iron (Fe) ≤0.0001 %
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR