Methylene blue trihydrate 95%, pure

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride trihydrate, C.I.52015, Methylthioninium chloride trihydrate, Basic blue 9, Swiss Blue, Methylene blue

126751000 126752500 126750010
ACRO126751000EA 124 EUR
ACRO126751000 ACRO126752500 ACRO126750010
Methylene blue trihydrate 95%, pure
Methylene blue trihydrate
Formula: C₁₆H₁₈ClN₃S·3H₂O
MW: 373.9 g/mol
Melting Pt: 100…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 7220-79-3
EINECS: 200-515-2

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥94 % (on dry substance)
Loss on drying ≤15 % (105°C)
Color scale ≤20 APHA
Acidity ≤0.001 meq/g
Alkalinity ≤0.002 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm
Halogenated compounds ≤0.001 % (as Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR