L(+)-Ornithine hydrochloride 99%

Supplier: Thermo Fisher Scientific

Synonyms: H-Orn-OH Hydrochloride, H-Orn-OH·HCl

129560250 129561000 129565000
ACRO129560250EA 37.9 EUR
ACRO129560250 ACRO129561000 ACRO129565000
L(+)-Ornithine hydrochloride 99%
L(+)-Ornithine hydrochloride
Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
Melting Pt: 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064562
CAS Number: 3184-13-2
EINECS: 221-678-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Solubility (5 % in 1 N HCl) Clear colorless
Specific optical rotation +21° to +24° (20°C, 589 nm) (c=5, 1 N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR