2-(Pyridin-3-yl)acetic acid hydrochloride 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Pyridyl-3-acetic acid hydrochloride, 3-Pyridylacetic acid hydrochloride

131960025 131960250
ACRO131960025EA 47.4 EUR
ACRO131960025 ACRO131960250
2-(Pyridin-3-yl)acetic acid hydrochloride 98%
2-(Pyridin-3-yl)acetic acid hydrochloride

CAS No.: 6419-36-9

Formula: C₇H₈ClNO₂
MW: 173.60 g/mol
Melting Pt: 160…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00012819
CAS Number: 6419-36-9

Order Now

Specification Test Results

Appearance White to light yellow to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
HPLC ≥96.0 %
Specific optical rotation -35° to -45° (20°C, 589 nm) (c=1, chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR