4,5,6,7-Tetrahydroisobenzofuran-1,3-dione 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-Tetrahydrophthalic anhydride

138220250 138220050
ACRO138220250EA 299 EUR
ACRO138220250 ACRO138220050
4,5,6,7-Tetrahydroisobenzofuran-1,3-dione 98%
4,5,6,7-Tetrahydroisobenzofuran-1,3-dione

Appearance: White to cream Crystalline powder

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00005917
CAS Number: 2426-02-0
UN: 2698
ADR: 8 III

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 74 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR