3,4,5-Trimethoxyaniline 97%

Supplier: Thermo Fisher Scientific
Warning

139900100 139900250
ACRO139900100EA 51.5 EUR
ACRO139900100 ACRO139900250
3,4,5-Trimethoxyaniline 97%
3,4,5-Trimethoxyaniline
Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008393
CAS Number: 24313-88-0
EINECS: 246-154-4
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Off-white to beige to light brown Powder
Infrared spectrum Conforms
Melting point 108 to 113 °C
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR