L(-)-Tyrosine 99+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH

140640010 140640050 140641000 140640025
ACRO140640010EA 446 EUR
ACRO140640010 ACRO140640050 ACRO140641000 ACRO140640025
L(-)-Tyrosine 99+%
L(-)-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351
Melting Pt: 290 °C (dec.)
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS Number: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Sulfated ash ≤0.1 %
Specific optical rotation -9.8° to -12.3° (20°C, 589 nm) (c=5, 1 N HCl)
Titration with HClO4 ≥99.0 % (on dried substance)
Chloride (Cl) ≤400 ppm
Sulfate (SO4) ≤300 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR