Methyl ethyl ketone 99+%, extra_pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

149670010 149670025 149670250 149670251
ACRO149670010EA 52.1 EUR
ACRO149670010 ACRO149670025 ACRO149670250 ACRO149670251
Methyl ethyl ketone 99+%, extra_pure
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: –87 °C
Density: 0.806 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3780 to 1.3800 (20°C, 589 nm)
Water ≤0.2 %
GC ≥99.0 %
Residue after evaporation ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR