Benzyl isocyanide 98%

Supplier: Thermo Fisher Scientific
Warning

158390010 158390050 158390250
ACRO158390010EA 73.1 EUR
ACRO158390010 ACRO158390050 ACRO158390250
Benzyl isocyanide 98%
Benzyl isocyanide
Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 105…106 °C (75 mmHg)
Density: 0.962 g/cm³
Flash Pt: 79 °C
Storage Temperature: Freezer
MDL Number: MFCD00000004
CAS Number: 10340-91-7
EINECS: 233-738-9
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear colorless to dark yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5220 (20°C, 589 nm)
GC ≥97.5 %
Iron (Fe) ≤0.003 %
Potassium (K) ≤0.02 %
Heavy metals (as Pb) ≤0.003 %
Insoluble matter ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR