2-Chlorobenzoic acid 98%

Supplier: Thermo Fisher Scientific
Warning

159240050 159241000 159245000 159240020
ACRO159240050EA 23.8 EUR
ACRO159240050 ACRO159241000 ACRO159245000 ACRO159240020
2-Chlorobenzoic acid 98%
2-Chlorobenzoic acid
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 285 °C (760 mmHg)
Melting Pt: 139…143 °C
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00002412
CAS Number: 118-91-2
EINECS: 204-285-4
Merck Index: 13,02143

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 139 to 143 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %
Copper (Cu) ≤5 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm
Selenium (Se) ≤10 ppm
Zinc (Zn) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR