4,6-Dichloropyrimidine 97%

Supplier: Thermo Fisher Scientific
Danger

163370050 163370250 163371000
ACRO163370050EA 52 EUR
ACRO163370050 ACRO163370250 ACRO163371000
4,6-Dichloropyrimidine 97%
4,6-Dichloropyrimidine
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 176 °C (760 mmHg)
Melting Pt: 63…68 °C
Density: 1.734
Storage Temperature: Ambient
MDL Number: MFCD00006109
CAS Number: 1193-21-1
EINECS: 214-770-2
UN: 3263
ADR: 8 II

Order Now

Specification Test Results

Appearance White to yellow to orange Powder or crystals or needles
Infrared spectrum Conforms
Melting point 64 to 70 °C
GC ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR