2-Methoxynaphthalene 98%

Supplier: Thermo Fisher Scientific

Synonyms: Methyl 2-naphthyl ether

164750050 164755000
ACRO164750050EA 21 EUR
ACRO164750050 ACRO164755000
2-Methoxynaphthalene 98%
2-Methoxynaphthalene
Formula: C₁₁H₁₀O
MW: 158.2 g/mol
Boiling Pt: 274
Melting Pt: 73…75 °C
MDL Number: MFCD00004061
CAS Number: 93-04-9
EINECS: 202-213-6
Merck Index: 12,06076

Order Now

Specification Test Results

Appearance White to yellow-brown Crystalline solid
Infrared spectrum Conforms
Melting point ≥71.5 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR