Methanol-[D₄] 99.5 atom % D NMR

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol

166350050 166350100 166350500
ACRO166350050EA 97.3 EUR
ACRO166350050 ACRO166350100 ACRO166350500
Methanol-[D₄] 99.5 atom % D NMR
Methanol-[D4]
Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.3
Melting Pt: –99 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
D-Enrichment ≥99.4 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR