Toluene-[D₈] 99+ atom % D NMR

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Phenylmethane-D8, Methylbenzene-D8, Toluene-[²H₈]

166390100 166390250
ACRO166390100EA 109 EUR
ACRO166390100 ACRO166390250
Toluene-[D₈] 99+ atom % D NMR
Toluene-[D8]
Formula: C₆D₅CD₃
MW: 100.08 g/mol
Boiling Pt: 110
Melting Pt: –84 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 4 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00044638
CAS Number: 2037-26-5
EINECS: 218-009-5
UN: 1294
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 %
D-Enrichment ≥99.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR