Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8% Biochemistry, analysis_for

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

167620010 167621000 167620051
ACRO167620010EA 188 EUR
ACRO167620010 ACRO167621000 ACRO167620051
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8% Biochemistry, analysis_for
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 219…220 °C (10 mmHg)
Melting Pt: >168.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Heavy metals ≤5 ppm
Sulfated ash ≤0.1 %
UV at 290 nm A: ≤0.2
pH 10.0 to 11.5 (5 % in water)
Titration with HClO4 ≥99.75 % (on dry substance)
Chloride (Cl) ≤5 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR