Tricine 98% Biochemistry

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine

172640250 172641000 172642500 172640010
ACRO172640250EA 31.4 EUR
ACRO172640250 ACRO172641000 ACRO172642500 ACRO172640010
Tricine 98% Biochemistry
Tricine
Formula: (HOCH₂)₃CNHCH₂CO₂H
MW: 179.17 g/mol
Melting Pt: 180…184 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
UV at 280 nm A: ≤0.02
pH 4.0 to 6.0 (1 % in water at 20°C)
Titration with HClO4 ≥97.5 % (On dried basis)
Heavy metals (as Pb) ≤5 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR