Deuterium chloride 20% w/w D2O 99+ atom % D NMR

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Hydrogen chloride-D1

175420500
ACRO175420500EA 129 EUR
ACRO175420500
Deuterium chloride 20% w/w D2O 99+ atom % D NMR
Deuterium chloride
Formula: DCl
MW: 37.45
Density: 1.2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00044642
CAS Number: 7698-05-7
EINECS: 231-715-8
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear colorless Solution
D-Enrichment ≥99.0 %
Concentration 20 to 22 % w/w
Ash ≤0.3 % (1000°C, 0.5 h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR