Polyvinyl alcohol, M.W. ∼ 13,000-23,000 87-89, hydrolysed

Supplier: Thermo Fisher Scientific

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

180300010 180300025 180301000
ACRO180300010EA 194 EUR
ACRO180300010 ACRO180300025 ACRO180301000
Polyvinyl alcohol, M.W. ∼ 13,000-23,000 87-89, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Melting Pt: 230…240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Appearance White to cream Granular powder
Infrared spectrum Conforms
Viscosity 3 to 3.7 cP (4 % in water at 20°C)
Degree of hydrolysis 87.0 to 89.0 mol %
Ash ≤1 % (as Na2O)
pH 4.5 to 6.5 (4 % in water)
Volatile matter ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR