Acetone-[D₆] 99.5 atom % D + 1% TMS for NMR spectroscopy

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Propanone-D6, DMK-D6, Hexadeuteroacetone, Dimethyl ketone-D6, Acetone-[²H₆], Acetone-d6

181160100 181160500
ACRO181160100EA 52.2 EUR
ACRO181160100 ACRO181160500
Acetone-[D₆] 99.5 atom % D + 1% TMS for NMR spectroscopy
Acetone-[D6]
Formula: D₃CC(O)CD₃
MW: 64.03 g/mol
Boiling Pt: 55.5
Melting Pt: –93 °C
Density: 0.87 g/cm³
Flash Pt: –20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00044635
CAS Number: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
D-Enrichment ≥99.4 %
HPLC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR