Methyl cellulose, 15 cP (2% H₂O)

Supplier: Thermo Fisher Scientific

Synonyms: Cellulose methyl ether

182311000 182312500 182310010
ACRO182311000EA 54 EUR
ACRO182311000 ACRO182312500 ACRO182310010
Methyl cellulose, 15 cP (2% H₂O)
Methyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Specification Test Results

Appearance White to yellowish-white to greyish-white Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Viscosity 12 to 18 cP (2 % in water at 20°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR