Poly(acrylic acid), M.W. ∼ 240,000 25% (w/w) in water

Supplier: Thermo Fisher Scientific

185010050 185012500 185015000
ACRO185010050EA 28 EUR
ACRO185010050 ACRO185012500 ACRO185015000
Poly(acrylic acid), M.W. ∼ 240,000 25% (w/w) in water
Poly(acrylic acid)

Appearance: Clear liquid

Formula: C₃H₄O₂
MW: 72.063 g/mol
Boiling Pt: 100 °C (760 mmHg)
Density: 1.09 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00084394
CAS Number: 9003-01-4

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Assay 23 to 27 % Solids
Molecular weight 240000 (approx)
HPLC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR