Zinc oxide 99.5+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

194890010 194890050
ACRO194890010EA 56.2 EUR
ACRO194890010 ACRO194890050
Zinc oxide 99.5+%
Zinc oxide
Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.6 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32

Order Now

Specification Test Results

Appearance White to faintly yellowish-white Powder
Titration Complexometric ≥99.50 %
Additional info Passes test
Particle size ≤0.01 % Sieve residue on 45 micron sieve
Residue after ignition ≤1 % (1 g, 500°C)
Specific surface area 3.5 to 9 m²/g
Alkalinity ≤0.3 ml/g (ml 0.1 N HCl/g)
Arsenic (As) ≤5 ppm
Cadmium (Cd) ≤10 ppm
Iron (Fe) ≤200 ppm
Lead (Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR