1-Fluoro-3-iodobenzene 99% Stabilised

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Fluoroiodobenzene

204480050 204480250
ACRO204480050EA 25.7 EUR
ACRO204480050 ACRO204480250
1-Fluoro-3-iodobenzene 99% Stabilised
1-Fluoro-3-iodobenzene
Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 77…78 °C (19 mmHg)
Melting Pt: –42 °C
Density: 1.89 g/cm³
Flash Pt: 67 °C
MDL Number: MFCD00001044
CAS Number: 1121-86-4
EINECS: 214-339-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5830 to 1.5860 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Copper


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR