Folic acid 97%, pure

Supplier: Thermo Fisher Scientific

Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

216630100 216630500 216632500
ACRO216630100EA 73.2 EUR
ACRO216630100 ACRO216630500 ACRO216632500
Folic acid 97%, pure
Folic acid
Formula: C₁₉H₁₉N₇O₆
MW: 441.41 g/mol
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00079305
CAS Number: 59-30-3
EINECS: 200-419-0

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Water ≤8.5 %
Sulfated ash ≤0.2 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR