Poly(styrene sulphonic acid)sodium salt, M.W. ∼ 70.000

Supplier: Thermo Fisher Scientific

Synonyms: Poly(sodium-p-styrenesulphonate)

222270050 222271000 222275000
ACRO222270050EA 40.5 EUR
ACRO222270050 ACRO222271000 ACRO222275000
Poly(styrene sulphonic acid)sodium salt, M.W. ∼ 70.000
Poly(styrene sulphonic acid)sodium salt

Appearance: White to tan Powder

Formula: [-CH₂CH(C₆H₄)SO₃Na)-]n
Storage Temperature: Ambient
MDL Number: MFCD00084449
CAS Number: 25704-18-1

Order Now

Specification Test Results

Appearance White to tan Powder
Infrared spectrum Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR