N-Acetyl-D-galactosamine 97%

Supplier: Thermo Fisher Scientific

Synonyms: 2-Acetamido-2-deoxy-D-galactose, D-N-Acetylgalactosamine, 2-Acetamino-2-deoxy-alpha-D-galactopyranose, GalNAc, N-Acetylchondrosamine

225850010 225855000
ACRO225850010EA 155 EUR
ACRO225850010 ACRO225855000
N-Acetyl-D-galactosamine 97%
N-Acetyl-D-galactosamine

CAS No.: 14215-68-0


Appearance: White to off-white Crystalline powder

Formula: C₈H₁₅NO₆
MW: 221.21 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Freezer
MDL Number: MFCD00065372
CAS Number: 14215-68-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤7.0 % (K.F.)
Solubility (10 % in water) Clear colorless
HPLC ≥96.0 % (ELSD)
Specific optical rotation +80° to +90° (20°C, 589 nm) (c=1, water) (after 5 h, on dry substance)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR