1-Butanol 99.5% analysis_for

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol

232080010 232080025 232080250
ACRO232080010EA 55.1 EUR
ACRO232080010 ACRO232080025 ACRO232080250
1-Butanol 99.5% analysis_for
1-Butanol
Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6 °C (760 mmHg)
Melting Pt: –89 °C
Density: 0.81 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 % (Coulometric)
GC ≥99.4 %
Color scale ≤10 APHA
Non-volatile matter ≤0.001 %
Substance colored by H2SO4 Passes test
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Cobalt (Co) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Free acid (C4H8O2) ≤0.01 %
Aldehyde (C4H8O) Passes test
Butyl ether ≤0.1 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR