4-Fluoro-2-nitroaniline 99%

Supplier: Thermo Fisher Scientific
Warning

233070100 233071000
ACRO233070100EA 31.8 EUR
ACRO233070100 ACRO233071000
4-Fluoro-2-nitroaniline 99%
4-Fluoro-2-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 92…95 °C
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00007830
CAS Number: 364-78-3
EINECS: 206-666-0

Order Now

Specification Test Results

Appearance Orange to brown Crystalline powder
Infrared spectrum Conforms
Melting point 92 to 95 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR