Silica gel, Pore size: 60 Å 0.035-0.070 mm Chromatography

Supplier: Thermo Fisher Scientific

240360010 240360050 240360300
ACRO240360010EA 102 EUR
ACRO240360010 ACRO240360050 ACRO240360300
Silica gel, Pore size: 60 Å 0.035-0.070 mm Chromatography
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9

Order Now

Specification Test Results

Appearance White Powder
Particle size ≤10 % (Oversize)
Specific gravity 0.46 to 0.55 kg/l (Typical)
pH 6.5 to 7.5 (5 % suspension in water) Typical
Residual water ≤6 % (160°C, 15 min)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR