2-(4-Isobutylphenyl)propanoic acid 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (±)-Ibuprofen, [(±)-2-(4-Isobutylphenyl)-propionic Acid, Ibuprofen

258610100 258610500 258610010
ACRO258610100EA 292 EUR
ACRO258610100 ACRO258610500 ACRO258610010
2-(4-Isobutylphenyl)propanoic acid 99%
2-(4-Isobutylphenyl)propanoic acid

CAS No.: 15687-27-1

Formula: C₁₃H₁₈O₂
MW: 206.3 g/mol
Boiling Pt: 157 °C (4 mmHg)
Melting Pt: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00010393
CAS Number: 15687-27-1

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 75 to 78 °C
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR