Zinc oxide 99.999% (trace metals basis)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

315790250 315791000
ACRO315790250EA 136 EUR
ACRO315790250 ACRO315791000
Zinc oxide 99.999% (trace metals basis)
Zinc oxide
Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.6 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32

Order Now

Specification Test Results

Appearance White to light yellow Powder and/or granules
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR