G418 disulphate Biochemistry

Supplier: Thermo Fisher Scientific
Danger

Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate

329400010 329400050
ACRO329400010EA 193 EUR
ACRO329400010 ACRO329400050
G418 disulphate Biochemistry
G418 disulphate
Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤10 %
Potency ≥650 µg/mg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR