4-Fluorophenethylamine 99%

Supplier: Thermo Fisher Scientific
Danger

350950050 350950250 350950500
ACRO350950050EA 28.7 EUR
ACRO350950050 ACRO350950250 ACRO350950500
4-Fluorophenethylamine 99%
4-Fluorophenethylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 50…52 °C (.1 mmHg)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00134208
CAS Number: 1583-88-6
EINECS: 216-435-6
UN: 2735
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5060 to 1.5080 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR