Pyridoxal hydrochloride 99%

Supplier: Thermo Fisher Scientific

Synonyms: 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl

352710050 352710010 352710250 352711000 352715000
ACRO352710050EA 75.5 EUR
ACRO352710050 ACRO352710010 ACRO352710250 ACRO352711000 ACRO352715000
Pyridoxal hydrochloride 99%
Pyridoxal hydrochloride
Formula: C₈H₁₀ClNO₃
MW: 203.62 g/mol
Melting Pt: 171…175 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012809
CAS Number: 65-22-5
EINECS: 200-602-5

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 171 to 175 °C (Dec)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR