4-Amino-3-bromopyridine 98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Bromo-4-pyridylamine, 3-Bromopyridin-4-amine

AC36062.1 AC36062.10 360620010 360620050
MAYBAC36062.1EA 0 EUR
MAYBAC36062.1 MAYBAC36062.10 ACRO360620010 ACRO360620050
4-Amino-3-bromopyridine 98%
4-Amino-3-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Boiling Pt: 244
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD02068297
CAS Number: 13534-98-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 71 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR