3-Aminophenylboronic acid pinacol ester 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester

367590010 367590050
ACRO367590010EA 69.6 EUR
ACRO367590010 ACRO367590050
3-Aminophenylboronic acid pinacol ester 97%
3-Aminophenylboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
Melting Pt: 89…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03453668
CAS Number: 210907-84-9

Order Now

Specification Test Results

Appearance Light brown Powder or crystals
Infrared spectrum Conforms
Melting point 89 to 94 °C
GC ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR