Chloroform-[D₁] 99.8 atom % D + 1% TMS stabilised for NMR spectroscopy

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D

368981000 368985000
ACRO368981000EA 98.2 EUR
ACRO368981000 ACRO368985000
Chloroform-[D₁] 99.8 atom % D + 1% TMS stabilised for NMR spectroscopy
Chloroform-[D1]
Formula: CDCl₃
MW: 120.38 g/mol
Boiling Pt: 60.8
Melting Pt: –64 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 %
Stabilizer silver foil
D-Enrichment ≥99.75 %
TMS 1 v/v%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR