Potassium iodide 99+% analysis_for

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Dipotassium diiodide, Kali iodide

373651000 373655000 373650025
ACRO373651000EA 49.2 EUR
ACRO373651000 ACRO373655000 ACRO373650025
Potassium iodide 99+% analysis_for
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330 °C (760 mmHg)
Melting Pt: 680 °C
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals
Titration Argentometric ≥99.0 % (on dry substance)
Loss on drying ≤0.2 % (150°C, 6 hrs)
pH 6.0 to 9.2 (5 % at 25°C)
Alkalinity ≤0.022 % (as KOH)
Iodate (IO3) ≤2 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm
Nitrogen compounds (as N) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR