2-Propanol 99.5% HPLC

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

383910010 383910025
ACRO383910010EA 79.3 EUR
ACRO383910010 ACRO383910025
2-Propanol 99.5% HPLC
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 81…83 °C (760 mmHg)
Melting Pt: –89.5 °C
Density: 0.785 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 %
GC ≥99.5 %
UV at 240 nm A: ≤0.05
Color scale ≤10 APHA
Residue after evaporation ≤2 ppm
Acidity/alkalinity ≤0.0001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR