N-Methylformamide 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Formmethylamide, Formic acid methylamide

414331000 414330010
ACRO414331000EA 28.8 EUR
ACRO414331000 ACRO414330010
N-Methylformamide 99%
N-Methylformamide
Formula: C₂H₅NO
MW: 59.07 g/mol
Boiling Pt: 199…201 °C (760 mmHg)
Melting Pt: –3.2 °C
Density: 1.003 g/cm³
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00003280
CAS Number: 123-39-7
EINECS: 204-624-6
Merck Index: 13,00061

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4340 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR