4-Fluoro-7-nitrobenzofurazan 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole, NBD-F

415450250 415450050
ACRO415450250EA 308 EUR
ACRO415450250 ACRO415450050
4-Fluoro-7-nitrobenzofurazan 98%
4-Fluoro-7-nitrobenzofurazan
Formula: C₆H₂FN₃O₃
MW: 183.1 g/mol
Melting Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010196
CAS Number: 29270-56-2

Order Now

Specification Test Results

Appearance Brown Crystalline chunks
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR