1-Butanol 99.5% ACS, meetSpec_Reagent Ph.Eur.

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol

423490010 423495000 423490025 423490100
ACRO423490010EA 64.3 EUR
ACRO423490010 ACRO423495000 ACRO423490025 ACRO423490100
1-Butanol 99.5% ACS, meetSpec_Reagent Ph.Eur.
1-Butanol
Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6 °C (760 mmHg)
Melting Pt: –89 °C
Density: 0.81 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
UN: 1120
ADR: 3 III
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 % (K.F.)
Boiling point 116 to 119 °C
Density (20/20 °C) 0.809 to 0.812
GC ≥99.4 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Carbonyl compounds ≤0.01 % (as butyraldehyde)
Titrable acid ≤0.0008 meq/g
Butyl ether ≤0.2 % (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR