Sodium tungstate dihydrate 99+% ACS

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate

424470050 424475000 424471000
ACRO424470050EA 26.5 EUR
ACRO424470050 ACRO424475000 ACRO424471000
Sodium tungstate dihydrate 99+% ACS
Sodium wolframate dihydrate
Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Melting Pt: 100
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2
EINECS: 236-743-4
Merck Index: 12,08847

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or flakes
Gravimetry 99.0 to 101.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤100 ppm
Molybdenum (Mo) ≤10 ppm
Heavy metals & Fe (as Pb) ≤0.001 %
Titrable free base ≤0.02 meq/g
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR