Toluene 99.5% ACS, meetSpec_Reagent Ph.Eur.

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

424550010 424555000 424550025 424550251
ACRO424550010EA 52 EUR
ACRO424550010 ACRO424555000 ACRO424550025 ACRO424550251
Toluene 99.5% ACS, meetSpec_Reagent Ph.Eur.
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 111 °C (760 mmHg)
Melting Pt: –95 °C
Density: 0.866 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
UN: 1294
ADR: 3 II
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.03 % (Coulometric)
Boiling point 109 to 111 °C
Density (20/20 °C) 0.865 to 0.870
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 passes test
Sulfur compounds (as S) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR