Copper(I)-2-thiophenecarboxylate 90% may contain ≈. 20% Copper(I) oxide

Supplier: Thermo Fisher Scientific
Danger

Synonyms: (2-Thiophenecarboxylato)copper(I), Copper(I) thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Copper(I) Salt, CuTC

430010010
ACRO430010010EA 92.3 EUR
ACRO430010010
Copper(I)-2-thiophenecarboxylate 90% may contain ≈. 20% Copper(I) oxide
Copper(I)-2-thiophenecarboxylate
Formula: C₅H₃CuO₂S
MW: 190.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02183524
CAS Number: 68986-76-5
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance Grey to red to brown Powder
Infrared spectrum Conforms
Assay 29.33 to 37.32 % (Cu) ICP-OES
Additional info may contain up to 0.3 mol % Copper(I)oxide
Elemental Analysis 14.80 to 18.83 % (S)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR