L(+)-Cysteine hydrochloride, anhydrous 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: H-Cys-OH.HCl, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride, L-Cysteine hydrochloride, (R)-(+)-Cysteine hydrochloride

434850010 434850250 434851000
ACRO434850010EA 1450 EUR
ACRO434850010 ACRO434850250 ACRO434851000
L(+)-Cysteine hydrochloride, anhydrous 98%
L(+)-Cysteine hydrochloride
Formula: HSCH₂CH(NH₂)COOH·HCl
MW: 157.62 g/mol
Melting Pt: 175…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00064553
CAS Number: 52-89-1
Merck Index: 13,02810

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis 8.67 to 9.11 % N
Specific optical rotation +6.5° to +9.5° (20°C, 589 nm) c=5 in 1N HCl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR