3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester 90%

Supplier: Thermo Fisher Scientific
Warning

445540010 445540050
ACRO445540010EA 303 EUR
ACRO445540010 ACRO445540050
3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester 90%
trans-3-(Tetrahydropyran-2-yloxy)propen-1-ylboronic acid pinacol ester
Formula: C₁₄H₂₅BO₄
MW: 268.16 g/mol
Boiling Pt: 79 °C (.1Torr)
Storage Temperature: Ambient
MDL Number: MFCD03788736
CAS Number: 642066-70-4

Order Now

Specification Test Results

Appearance Colorless Oily liquid
GC ≥88.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR