Tetramethylsilane ACS for NMR spectroscopy

Supplier: Thermo Fisher Scientific
Danger

Synonyms: TMS

445900250 445901000
ACRO445900250EA 41.4 EUR
ACRO445900250 ACRO445901000
Tetramethylsilane ACS for NMR spectroscopy
Tetramethylsilane
Formula: C₄H₁₂Si
MW: 88.22 g/mol
Boiling Pt: 26…28 °C (760 mmHg)
Melting Pt: –99 °C
Density: 0.64 g/cm³
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008274
CAS Number: 75-76-3
UN: 2749
ADR: 3 I

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Residue after evaporation ≤0.05 %
Suitability (for 1H-NMR position ref.) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR